missing translation for 'onlineSavingsMsg'
Learn More

GRB2 Mouse anti-Human, Mouse, Rat, CoraLite 594, Clone: 1C5D11, Proteintech

Mouse Monoclonal Antibody

1696.32 NOK - 5264.45 NOK

Specifications

Antigen GRB2
Clone 1C5D11
Concentration 1000 μg/mL
Applications Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17711439
View Documents
Proteintech
CL594-66880-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
17701439
View Documents
Proteintech
CL594-66880-100UL
100 μL
5264.45 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GRB2 Monoclonal antibody specifically detects GRB2 in Human, Mouse, Rat samples. It is validated for IF
Specifications

Specifications

GRB2
1000 μg/mL
Monoclonal
Liquid
Human, Mouse, Rat
2885
Primary
Store at -20°C
1C5D11
Immunofluorescence
CoraLite 594
Mouse
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
IgG1
Protein G purification
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.