Learn More
Thermo Scientific™ Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Improve your AAS graphite furnace performance using Thermo Scientific™ application-specific graphite cuvettes.
12970.00 NOK
Specifications
Type Graphite Furnace Cuvette
For Use With (Equipment) GFS33 or GFS35 Furnaces
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11517943
View Documents
Thermo Scientific™
942349020101
Omega Cuvette with Integral Platform
12970.00 NOK
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
Description
Description
Discover different types of graphite furnace cuvettes suitable for any analytical application using the Thermo Scientific™ GFS33/GFS35/GFS35Z Graphite Furnace and Autosampler modules
Improve instrument performance with the right selection of graphite cuvettesWhile ridged cuvettes are supplied as standard with Thermo Scientific GFS33/GFS35/GFS35Z Graphite Furnaces, additional application-specific cuvettes improve performance for more complex applications:
  • Extended Lifetime Cuvettes (ELCs) can be used longer and provide reproducible performance over several thousand firings
  • Ridged Cuvettes (in pyrolytically-coated and uncoated forms) help maintain difficult samples at the optimum temperature for efficient atomization
  • Omega Cuvettes with integral platforms provide the most interference-free conditions (often in combination with matrix modification) for samples with complex matrices
Specifications
Specifications
Graphite Furnace Cuvette
GFS33 or GFS35 Furnaces