Learn More
Kartell Graduated Micro Test Tubes
Graduated Micro Test Tubes ideal for sample analysis.
Brand:  Kartell 0027913
 View more versions of this product
Product Code. 18028041
  • 2540.00 NOK / Pack of 10000
Explore more special offers
Description
Description
  • Graduated Micro Test Tubes ideal for sample analysis
  • Flat cap with writing area
  • Ideal for samples analysis
  • Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to RCF 11,000
  • Autoclavable Up to 121°C for 20 minutes with open cap
Specifications
Specifications
Autoclavable
Pink
Non-sterile
Attached Snap Cap
Conical
Microcentrifuge Tube
1.5 mL
Polypropylene (Virgin)
Yes
Cylindrical
11,000 g