Learn More
Corning™ Gosselin™ Self Standing Tubes
Applications: Environmental and Water Testing, Food and Beverage Testing, Pharmaceutical Quality Control and Life Sciences
Brand:  Corning™ TP10-02
Product Code. 12367547
  • 1241.70 NOK / Pack of 1000
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Description
  • Polypropylene (PP) 
  • Self-standing tubes
  • Sterile or nonsterile
Note: Cap is not included; sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mL
250/Pk.
Round Base
No
Polypropylene
1000/Cs.
Round Base