Learn More
Gold powder, spherical, APS 0.5-0.8 micron, 99.96+% (metals basis), Thermo Scientific Chemicals Product Code.: 11425460
Gold powder, spherical, APS 0.5-0.8 micron, 99.96+% (metals basis), Thermo Scientific Chemicals
Product Code. 11425460
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
Product Code. 11425460
Brand: Thermo Scientific Alfa Aesar 044636.MD
Request Bulk or Custom Format
Form: S.A. 0.45-0.70 m2/g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
Specifications
Color Golden
Appearance Gold colored powder
Assay Percent Range >99.96% (Metals basis)
Quantity 250 mg
Odor Odorless
Form Spherical Powder
Chemical Name or Material Gold
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX29K059 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number044636.06,044636.03,044636.MD
Lot NumberU25H006 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog Number044640.02,044636.06,044636.03,044640.04,044636.MD
Lot NumberW28H021 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog Number044640.02,044636.06,044636.03,044640.04,044636.MD
Lot NumberY08I022 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog Number044640.02,044636.06,044636.03,044640.04,044636.MD
Lot NumberN02H026 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044640.02,044636.06,044636.03,044640.04,044636.MD
Product Title
Gold powder, spherical, APS 0.5-0.8 micron, 99.96+% (metals basis), Thermo Scientific Chemicals >