Learn More
Gold foil, 0.5mm (0.02in) thick, Premion™, 99.9985% (metals basis) Product Code.: 11385849
Gold foil, 0.5mm (0.02in) thick, Premion™, 99.9985% (metals basis)
Product Code. 11385849
50 x 50 mm, Each
Click to view available options
Quantity:
50 x 50 mm
Unit Size:
Each
Product Code. 11385849
Brand: Thermo Scientific Alfa Aesar 011389.FI
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
CAS: 7440-57-5
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7440-57-5
Quantity 50 x 50 mm
Percent Purity 99.9985%
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberT08K065 Certificate TypeCertificate of Analysis Date30/07/2024 Product Code011389.FI,011389.FF
Lot NumberN13K048 Certificate TypeCertificate of Analysis Date23/02/2024 Product Code011389.FI,011389.FF
Lot NumberP01J018 Certificate TypeCertificate of Analysis Date09/05/2023 Product Code011389.FI
Lot NumberY02I022 Certificate TypeCertificate of Analysis Date15/11/2022 Product Code011389.FI,011389.FF
Lot NumberW26D010 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code011389.FI,011389.FF
Product Content Correction
Product Title
Gold foil, 0.5mm (0.02in) thick, Premion™, 99.9985% (metals basis) >