Learn More
3M™ Goggle Gear™ Safety Goggles, Anti-Fog / Anti-Scratch Coating (K&N) Product Code.: 17237680
Only undefined left
Add to basket
3M™ Goggle Gear™ Safety Goggles, Anti-Fog / Anti-Scratch Coating (K&N)
Product Code. 17237680
Gray, Pack of 10
Click to view available options
Lens Tint:
Gray
Transparent
Unit Size:
Pack of 10
Product Code. 17237680
Brand: 3M™ 7100185290
Only undefined left
Add to basket
Only undefined left
Add to basket
Goggle Gear™ Anti-Fog / Anti-Scratch Coating (K&N)
  • Modern slim-line and lightweight design for an excellent fit
  • Adjustable Neoprene headband for easy cleaning and sterilisation
  • Pivoting ratchet for convenient personal adjustment
  • 3M™ Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties
  • Coating withstands washing, soaking in bleach or using alcohol wipes without losing performance
  • Special 3M™ Goggle Gear prescription insert available
  • Offers excellent protection against UV radiation
  • Lens Marking: 2C-1.2 3M 1 BT KN
  • Frame Marking: 3M EN 166 3 4 BT CE
  • Protection against large dust particles (3)
  • Protection against liquid droplets (4)
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Gray
Frame Color Gray and Red
Model GG502SGAF-EU
Quantity 10/Cs.
Lens Material Polycarbonate
Frame Type Goggle
Product Content Correction
Product Title
3M™ Goggle Gear™ Safety Goggles, Anti-Fog / Anti-Scratch Coating (K&N) >