Learn More
GNE Mouse anti-Human, Unconjugated, Clone: 2A11E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67649-1-IG-20UL
 View more versions of this product
Product Code. 16860396
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The GNE antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNE. This antibody recognizes human antigen. The GNE antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
GNE
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot
2A11E10
Unconjugated
20 μL
10020
IgG1