Learn More
GNE Mouse anti-Human, Unconjugated, Clone: 2A11E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67649-1-IG-150UL
This item is not returnable.
View return policy
Description
The GNE antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNE. This antibody recognizes human antigen. The GNE antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
GNE | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot | |
2A11E10 | |
Unconjugated | |
150 μL | |
10020 | |
IgG1 |