missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMF-beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10690-1-AP-150UL
This item is not returnable.
View return policy
Description
This protein, which is phosphorylated following phorbol ester stimulation, is important for the nervous system. It causes brain cell differentiation, stimulates neural regeneration and inhibits tumor cell proliferation. Overexpression of GMF in astrocytes has been shown to enhance brain-derived neurotrohic factor (BDNF) production. GMF expression is increased by exercise, and the protein is crucial for exercise-induction of BDNF. Through BDNF production, GMF appears to play a role in neuroprotection. In thymoma, T-cell development is maintained by GMF; being produced by the tumor cells.Specifications
GMF-beta | |
Polyclonal | |
Unconjugated | |
GMFB | |
Glia maturation factor beta, glia maturation factor, beta, GMF, GMF beta, GMFB, GMF-beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2764, 63985, 81661 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P60983, Q63228, Q9CQI3 | |
GMFB | |
GMF-beta Fusion Protein Ag1104 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction