Learn More
Multiroir Controlec™ GM Sectioned Drawer Inserts
Fits securely in the tray of a GM Drawer Cabinet. Multiroir Controlec™ GM Sectioned Drawer Inserts are thermoformed honeycomb grid pieces that customize the plastic drawer with the size and number of compartments needed for sectioned storage.
174.00 NOK - 176.00 NOK
Specifications
Material | Plastic |
---|---|
Product Type | Sectioned Drawer Insert |
For Use With (Application) | Sectioned storage |
Height (Metric) | 40 mm |
Color | White |
Product Code | Brand | Array | Width (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Array | Width (Metric) | Length (Metric) | Price | Quantity & Availability | |||
11774571
|
Multiroir Controlec™
32154 |
3 x 4 | 79 mm (compartment) | 77 mm (compartment) |
174.00 NOK
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11784571
|
Multiroir Controlec™
32155 |
4 x 4 | 55 mm (compartment) | 77 mm (compartment) |
176.00 NOK
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
Description
GM Drawer Cabinets are a block of six colored tray-drawers to suit a variety of sorted storage needs.
- Solid polypropylene construction
- Color coded trays / drawers
- Transportable and stackable
- Drawers can be equipped with a variety of honeycomb grids
- Drawer/Insert Size: 340 x 250 x 40mm
Specifications
Plastic | |
Sectioned storage | |
White | |
77 mm (compartment) |
Sectioned Drawer Insert | |
40 mm | |
GM Drawer Cabinets |