missing translation for 'onlineSavingsMsg'
Learn More
Glycine tert-butyl ester hydrochloride, 99% Product Code.: 11353956

Glycine tert-butyl ester hydrochloride, 99%

Product Code. 11353956
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11353956

Brand: Thermo Scientific Alfa Aesar L17116.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27532-96-3 | C6H14NO2 | 132.18 g/mol

Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

Solubility
Soluble in methanol and water

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27532-96-3
Molecular Formula C6H14NO2
Molecular Weight (g/mol) 132.18
MDL Number MFCD00058255
InChI Key SJMDMGHPMLKLHQ-UHFFFAOYSA-O
Synonym h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride
PubChem CID 2725036
IUPAC Name tert-butyl 2-aminoacetate;hydrochloride
SMILES CC(C)(C)OC(=O)C[NH3+]

Specifications

Melting Point ∽140°C (decomposition)
Beilstein 3989048
Solubility Information Soluble in methanol and water
Quantity 1 g
Formula Weight 167.64
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material Glycine tert-butyl ester hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10249612 Certificate TypeCertificate of Analysis Date06/03/2024 Product CodeL17116.03,L17116.06
Lot Number10211344 Certificate TypeCertificate of Analysis Date22/11/2023 Product CodeL17116.03,L17116.06
Lot Number10247006 Certificate TypeCertificate of Analysis Date20/06/2023 Product CodeL17116.03,L17116,L17116.06
Lot Number10205300 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeL17116.03,L17116.06
Lot Number61500179 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL17116.03,L17116.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycine tert-butyl ester hydrochloride, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.