Learn More
Glycine benzyl ester p-toluenesulfonate salt, 99%
Description
Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.
Solubility
Soluble in methanol. (0.1 g/mL)
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Specifications
Specifications
Melting Point | 132°C to 134°C |
Linear Formula | NH2CH2COOCH2C6H5·CH3C6H4SO3H |
Quantity | 5 g |
Beilstein | 3600116 |
Solubility Information | Soluble in methanol. (0.1g/mL) |
Formula Weight | 337.39 |
Percent Purity | 99% |
Chemical Name or Material | Glycine benzyl ester p-toluenesulfonate salt |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.