Learn More
Scharlab GLYCERIN 86% PA 1L
Brand: Scharlab GL00231000
GLYCERIN 86% PA 1L
This item is not returnable.
View return policy
Product Content Correction
Product Title
Scharlab GLYCERIN 86% PA 1L >