Learn More
MP Biomedicals™ Gly-Phe-7-Amino-4-Trifluoromethylcoumarin
Gly-Phe-7-Amino-4-Trifluoromethylcoumarin, White powder
Brand:  MP Biomedicals™ 03AFC03310
 View more versions of this product
Product Code. 11439795
  • 2885.00 NOK / 10mg
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gly-Phe-AFC is a fluorogenic, cell-permeant peptide substrate, used in Cytotoxicity testing.
  • In vitro cytotoxicity testing has become an integral aspect of drug discovery because it is a convenient, costeffective, and predictive means of characterizing the toxic potential of new chemical entities.
  • It is also used as substrate for measuring Dipeptidyl peptidase1 (DPP1) activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Cell Migration Analysis; Cell Signaling Analysis
433.38 ; 574 (Including TFA salt)
Keep cool and dry
Powder
10mg
Proteins, Enzymes & Peptides