Learn More
Tocris Bioscience™ GLP-1 (7-37)
Endogenous bioactive GLP-1 receptor ligand
Brand: Tocris Bioscience™ 5374/1
This item is not returnable.
View return policy
Description
GLP-1 (7-37) is a endogenous GLP-1 receptor ligand; bioactive and truncated form of GLP-1; insulinotropic hormone.Specifications
0.95 | |
Store at -20°C | |
Observed M.W.: 3355.71 | |
C151H228N40O47 |
106612-94-6 | |
Glucagon-Like Peptide 1 Receptor Agonists | |
1 mg |