Learn More
Karl Hecht™ Glass Vials with Plastic Snap-Cap
With a minimum order quantity of 1 box.
Brand:  Karl Hecht™ 42785016
 View more versions of this product
Product Code. 10509398
  • 1505.00 NOK / Pack of 200
Only null left
Estimated Shipment: 08-07-2025
Description
  • Flat bottom
  • With rolled rim
  • Available in a range of capacities
Specifications
Specifications
Natural
Approx. 30 mm
∼25 mL
Snap Cap
Rolled rim
Glass
Plastic
200/Pk.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Karl Hecht™ Glass Vials with Plastic Snap-Cap > Capacity: 25mL