Learn More
Poulten Selfe™ Glass Ubbelohde
Suitable for transparent liquids
Brand: Poulten Selfe™ 1643/06
This item is not returnable.
View return policy
Description
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.Specifications
Ubbelohde Viscometer | |
Yes | |
1B |
10 to 50 cSt | |
Glass |
Product Content Correction
Product Title
Poulten Selfe™ Glass Ubbelohde >