Learn More
Poulten Selfe™ Glass Ubbelohde
Suitable for transparent liquids
Brand:  Poulten Selfe™ 1643/11
 View more versions of this product
Product Code. 13111073
  • 2560.00 NOK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.
Specifications
Specifications
Ubbelohde Viscometer
Yes
3C
600 to 3000 cSt
Glass
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Poulten Selfe™ Glass Ubbelohde >