Learn More
Bochem™ Glass Tube Cutters
Includes hard metal wheel. Bochem Glass Tube Cutters accommodates a range of diameters.
Brand:  Bochem™ 12210
 View more versions of this product
Product Code. 10644691
  • 279.00 NOK / Each
Only null left
Estimated Shipment: 31-07-2025
Description
  • With hard metal wheel
Specifications
Specifications
MS-Ni
40 mm
Glass Tubing Cutter
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Glass Tube Cutters > Diameter: 40mm