Learn More
Duran™ Glass Test Tubes Straight Rim
Thick-walled, straight-rim test tubes. Duran™ Glass Test Tubes Straight Rim are mechanically resistant, yet still retain good thermal shock resistance.
672.00 NOK
Specifications
Rimmed Straight Rim
Product Type Test Tube
View More Specs
Products 1
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
10463151
View Documents
Duran™
26 131 06 03
100 mm
672.00 NOK
Pack of 100
Estimated Shipment: 24-04-2025
Description
  • Available in a variety of diameters, heights and volumes (2.0-100mL)
  • Durable Duran™ borosilicate glass construction
Specifications
Specifications
Straight Rim
Test Tube
Documents
Product Certifications
Product Content Correction
Product Title
Duran™ Glass Test Tubes Straight Rim