Learn More
Duran™ Glass Stoppers
Manufactured from Borosilicate glass. DURAN™ Glass Stoppers are semi-hollow.
Brand:  Duran™ 286114804
1819.25 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 15901521
  • 1915.00 NOK / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • DURAN™ cap
  • Flat head
  • Semi-hollow
  • Made of Borosilicate glass
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Duran™ Glass Stoppers > Cone Size: NS 12/21; Shape: Hexagonal; Material: Glass; Tip Style: Pointed