Learn More
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End
Facilitates error-free capillary filling in the laboratory. Hirschmann Instruments™ Disposable Micro Capillary Pipettes, End-to-End are manufactured from durable DURAN™ glass.
1400.00 NOK - 1625.00 NOK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
11787663
|
Hirschmann™
9000203 |
3 ÎĽL |
1620.00 NOK
Pack of 1000 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
11797663
|
Hirschmann™
9000204 |
4 ÎĽL |
1620.00 NOK
Pack of 1000 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
11767663
|
Hirschmann™
9000201 |
1 ÎĽL |
1620.00 NOK
Pack of 1000 |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
11777663
|
Hirschmann™
9000202 |
2 ÎĽL |
1620.00 NOK
Pack of 1000 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
11707673
|
Hirschmann™
9000205 |
5 ÎĽL |
1620.00 NOK
Pack of 1000 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
11730055
|
Hirschmann™
9000299 |
100 ÎĽL |
1620.00 NOK
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10720841
|
Hirschmann™
9001166 |
6.66 ÎĽL |
1400.00 NOK
Pack of 1000 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
11717673
|
Hirschmann™
9000210 |
10 ÎĽL |
1620.00 NOK
Pack of 1000 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
11772533
|
Hirschmann™
9000220 |
20 ÎĽL |
1620.00 NOK
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10582501
|
Hirschmann™
9000225 |
25 ÎĽL |
1625.00 NOK
Pack of 1000 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
10480771
|
Hirschmann™
9000240 |
40 ÎĽL |
1620.00 NOK
Pack of 1000 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
11797673
|
Hirschmann™
9000250 |
50 ÎĽL |
1620.00 NOK
Pack of 1000 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
Description
- End-to-end, with minicaps
- Conformity certified
- Available either heparinized or non-heparinized
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
0.5% | |
Minicap Capillary Pipette | |
Single-use |
Product Title
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End