Learn More
Poulten Selfe™ Glass Routine Viscometer
Suitable for measuring transparent liquids
Brand:  Poulten Selfe™ 1634/01
 View more versions of this product
Product Code. 13141073
  • 2500.00 NOK / Each
Only null left
Estimated Shipment: 28-07-2025
Description
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids, nominal overall length 250mm. Approximate sample size 7mL. Calibration data at 40°C with the constant, C, stated at 40°C and 100°C. With NAMAS certification of calibration.
Specifications
Specifications
Routine Viscometer
Yes
25
0.5 to 2 cSt
Glass
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Poulten Selfe™ Glass Routine Viscometer > nominal constant 0.002 0.5 to 2 viscosity range size 25 Poulten Selfe and Lee