Learn More
IKA™ Glass Receiving Flask
Brand: IKA™ 0003742200
This item is not returnable.
View return policy
Description
Specifications
Glass | |
Round | |
Round |
Receiving Flask | |
1 Pack | |
100 mL |
Product Content Correction
Product Title
IKA™ Glass Receiving Flask > Capacity: 100mL