Specifications
Length (Metric) | 76 mm |
Width (Metric) | 60 mm |
Type | Glass Plate |
Well Count | 12 |
Color | Transparent |
Diameter (Metric) | 16 mm |
Quantity | 1 Pack |
Dimensions (L x W) | 76 x 60 mm |
Product Content Correction
Product Title
Hecht Karl™ Glass Plate >