Learn More
Karl Hecht™ Glass Pasteur Pipettes
Disposable pipettes with a uniform shape for very precise applications in the laboratory.
438.00 NOK - 451.00 NOK
Specifications
Diameter (Metric) Outer | 7 mm |
---|---|
Material | Soda Lime Glass |
For Use With (Application) | Pipetting |
Quantity | 250/Box, 1000/Carton |
Tip Style | Long Tapered |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10491122
|
Karl Hecht™
40567091 |
150 mm |
451.00 NOK
Pack of 250 |
Estimated Shipment: 23-09-2025
Log in to see stock. |
||||||
10708472
|
Karl Hecht™
40567092 |
230 mm |
438.00 NOK
Pack of 250 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
Description
- Disposable
- Acc. to ISO 7712
- Manufactured of soda glass
- Long tapered tip
- With constriction in the upper part for cotton plug
- Capillary tip inside diameter of approx. 1mm, body approx. 90 x 7mm
- Short form and long form available
- 250 pieces/box, 1000 pieces/carton
- With CE marking
- 100 pieces/tube; 10 tubes/box
Specifications
7 mm | |
Pipetting | |
Long Tapered |
Soda Lime Glass | |
250/Box, 1000/Carton | |
Single-use |
Product Title
Karl Hecht™ Glass Pasteur Pipettes