Learn More
WTW™ Glass Overflow Measuring Flask
Replacement sample bottles and other glassware as well as useful accessories for the expansion of application of measuring systems for different parameters.WTW offers a large assortment of glass ware and other products to replace, supplement and to extend your applications.
Brand: WTW™ 209090
772.12 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
432 mL | |
BOD | |
Glass |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
WTW™ Glass Overflow Measuring Flask > Capacity: 432mL