Learn More
Kimble Chase™ Glass Funnel
Brand:  Kimble Chase™ 953701-0325
Product Code. 11753459
  • 3300.00 NOK / Each
Only null left
Estimated Shipment: 18-09-2025
Specifications
300 mL
Glass
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kimble Chase™ Glass Funnel > Capacity: 300mL