missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Glass Filter Plugs without Tube

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 07205120

Additional Details : Weight : 0.25000kg

Product Code. 10872611

  • 414.46 NOK / Each
Estimated Shipment: 03-10-2024
to see stock.
Add to basket

Description

Description

  • Filter plugs
  • Cylindrical
  • With tube (dia. 8 mm x length 210 mm)
  • Material: borosilicate glass 3.3
Specifications

Specifications

Borosilicate Glass (3.3)
1
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers