Learn More
Brady™ Glass-Filled Nylon Small Blocking Arm
Glass-Filled Nylon Small Blocking Arm for use with small Universal Valve Lockout.
Brand: Brady™ 065402
484.50 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Description
- Blocking arm for use with small Universal Valve Lockout.
Specifications
Blocking Arm | |
Red | |
153.7 x 157.48 mm |
UNIVERSAL BALL VALVE L/O SML ARM ADAPTER | |
Small Blocking Arm | |
1 Ea. |
Product Content Correction
Product Title
Brady™ Glass-Filled Nylon Small Blocking Arm > Glass-Filled Nylon Small Blocking Arm; Dimensions (W x H): 157.50 x 153.70 mm