Learn More
DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals
Kimble™ Glass-filled Nylon Connectors with PTFE Seals are threaded connectors used for coupling two items with a GPI finish.
299.00 NOK - 405.00 NOK
Specifications
Material | Nylon |
---|---|
Product Type | Connector |
For Use With (Application) | To connect standard rotary evaporator traps to vials |
Includes | PTFE seals |
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
11892933
|
DWK Life Sciences
747205-1820 |
18-400 to 20-400 |
395.00 NOK
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||||
11882933
|
DWK Life Sciences
747205-1520 |
15-425 to 20-400 |
385.00 NOK
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||||
17751088
|
DWK Life Sciences
747205-1320 |
13-425 to 20-400 |
299.00 NOK
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||||
11802943
|
DWK Life Sciences
747205-2020 |
20-400 to 20-400 |
405.00 NOK
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||||
Description
- Connectors have PTFE seals and FKM O-Rings
- Blue glass-filled nylon
- For use to 150°C
Specifications
Nylon | |
Connector | |
To connect standard rotary evaporator traps to vials | |
PTFE seals |
Product Title
DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals