Learn More
BRAND™ Glass Discharge Tip
Shop All Brand Products
Click to view available options
For Use With (Equipment):
Automatic Burette
Unit Size:
Pack of 10
Description
- Clear AR-GLAS™ construction
- For automatic burette
- Schilling pattern
- Includes silicone tubing
Specifications
Specifications
Color | Clear |
For Use With (Equipment) | Automatic Burette |
Material | Glass |
Includes | Silicone tubing |
Type | Discharge Tip |
Product Content Correction
Product Title
BRAND™ Glass Discharge Tip >