Learn More
Eppendorf™ Glass Cuvette
Designed for a variety of applications
Brand: Eppendorf™ 4052102001
6480.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Optical glass composition
- Available in a variety of models and sizes
Specifications
Cuvette | |
Glass | |
Spectrophotometer | |
Spectroscopy |
Model: 4052, flow-through | |
1 Each | |
10 mm |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
Eppendorf™ Glass Cuvette > Model: 4052