missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Hecht™ Glass Centrifuge Tubes, Conical
Clear, glass centrifuge tubes provide superior visibility. Karl Hecht™ Glass Centrifuge Tubes, Conical are available graduated/ungraduated, rimmed/rimless, and constructed of AR™ soda glass or Duran™ borosilicate.
Brand: Karl Hecht™ 40942001
This item is not returnable.
View return policy
Description
- Conical bottom.
- Graduated or ungraduated.
- Rimmed or rimless.
- AR™ soda glass, or Duran™ borosilicate.
Specifications
10 to 15 mL | |
Centrifuge | |
112 mm | |
17 mm | |
Rimmed Centrifuge Tube |
Soda Lime Glass | |
100/Box | |
Conical | |
10 mL, 1/10 with mark at 15 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Hecht™ Glass Centrifuge Tubes, Conical > Graduations: 10mL, 1/10 with mark at 15mL, Length: 112mm, Material: AR™ Soda Glass,Type: Rimmed
Spot an opportunity for improvement?Share a Content Correction