Learn More
Alpha Packaging™ Round Glass Narrow Bottles
Use Alpha Packaging™ Round Glass Narrow Bottles for bottling needs as your products dictate.
621.00 NOK - 22643.20 NOK
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Price | Quantity & Availability | |||||
30408798
|
Alpha Packaging™
1000741 |
100 mL | Amber |
22643.20 NOK
Pack of 3168 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10014544
|
Alpha Packaging™
232076 |
250 mL | Amber |
621.00 NOK
Pack of 51 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
30408788
|
Alpha Packaging™
1000736 |
250 mL | Clear |
16158.16 NOK
Pack of 1428 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30408782
|
Alpha Packaging™
1000834 |
500 mL | Amber |
16775.43 NOK
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30408789
|
Alpha Packaging™
1000742 |
1000 mL | Clear |
12772.66 NOK
Pack of 420 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- DIN 28 certification
- Amber or clear
- Round glass with narrow neck
Specifications
DIN 28 | |
Glass |
Product Title
Alpha Packaging™ Round Glass Narrow Bottles