Learn More
Sartorius™ Glass Bell Jar
Brand:  Sartorius™ YAW03
 View more versions of this product
Product Code. 10022191
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Specifications
Bell Jar
Bell
1 Each
Glass
2kg to 5kg Weights
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Glass Bell Jar > For Use with 2kg to 5kg Weights