Learn More
Sartorius™ Glass Bell Jar
Brand:  Sartorius™ YAW02
 View more versions of this product
Product Code. 10012191
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Specifications
Bell Jar
Bell
1 Each
Glass
100g to 1kg Weights
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Glass Bell Jar > For Use with 100g to 1kg Weights