Learn More
Glass Balls, Pure, 3.7-4.1mm, Undrilled, Fisher Chemical™ Product Code.: 10375750
Glass Balls, Pure, 3.7-4.1mm, Undrilled, Fisher Chemical™
Product Code. 10375750 Shop All Fisher Chemical Products
250 g, 250g
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
Product Code. 10375750
Brand: Fisher Chemical G/0300/50
Request Bulk or Custom Format
Glass Balls, Pure, 3.7-4.1mm, Undrilled, Fisher Chemical, Quantity: 500g, Packaging: Plastic powder jar, MDL Number: 81645
Chemical Identifiers
MDL Number 81645
Specifications
Packaging Plastic powder jar
Quantity 250 g
Product Content Correction
Product Title
Glass Balls, Pure, 3.7-4.1mm, Undrilled, Fisher Chemical™ >