Learn More
Edmund Buehler™ Glass Ball
Brand:  Edmund Buehler™ 0001 067
Product Code. 10738092
  • 459.00 NOK / 1kg
Only null left
Estimated Shipment: 22-07-2025
Specifications
Ball
Glass
1 mm
1 kg
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Edmund Buehler™ Glass Ball > Diameter: 1mm