missing translation for 'onlineSavingsMsg'
Learn More

Ginsenoside Rb1 Reference MP Biomedicals

Product Code. 11436861
Click to view available options
Quantity:
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 41753-43-9
Molecular Formula C54H92O23
Quantity 5 mg
InChI Key GZYPWOGIYAIIPV-VOXLZBFFSA-N
SMILES CC(=CCCC(C)(C1CCC2(C1C(CC3C2(CCC4C3(CCC(C4(C)C)OC5C(C(C(C(O5)CO)O)O)OC6C(C(C(C(O6)CO)O)O)O)C)C)O)C)OC7C(C(C(C(O7)COC8C(C(C(C(O8)CO)O)O)O)O)O)O)C
IUPAC Name (2R,3R,4S,5S,6R)-2-[[(2R,3S,4S,5R,6S)-6-[(2S)-2-[(3R,8R,10R,12S,13S,14S,17S)-3-[(2R,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-12-hydroxy-4,4,8,10,14-pentamethyl-2,3,5,6,7,
Molecular Weight (g/mol) 1109.307
PubChem CID 131676146
Formula Weight 1109.3
Grade Reference
Chemical Name or Material Ginsenoside Rb1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.