Learn More
Ginkgolide A, MedChemExpress
MedChemExpress Ginkgolide A (BN-52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist.
Brand:  Medchem Express HY-B0355
Encompass_Preferred
 View more versions of this product
Product Code. 16418458
  • 647.96 NOK / 10mg
Explore more special offers
Description
Description
Ginkgolide A (BN-52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist.
Specifications
Specifications
Ginkgolide A
White
C20H24O9
BN-52020
O=C1[C@@H](C)[C@]2(O)[C@@]3(C4=O)C(C5([C@@H](O)C(O6)=O)C6O3)([C@H](O4)C[C@H]5C(C)(C)C)C[C@]2([H])O1
408.4
Research
COVID-19-immunoregulation
15291-75-5
Research
10 mg
DMSO : 100 mg/mL (244.86 mM; Need ultrasonic)
408.4
98.0%
Solid