Learn More
Fisherbrand™ Hotplate Stirrer Clamp
For Use With: Isotemp Hotplates, Stirrers and Stirring Hotplates
Brand:  Fisherbrand™ CIC0001833
372.74 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15954520
  • 432.60 NOK / Each
In Stock
to see stock.
Explore more special offers
Description
Clamp and clamp holder for use with Isotemp hotplates, stirrers and stirring hotplates
  • Clamp
  • Clamp holder
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Attaches to supporting rod
Isotemp Hotplates, Stirrers and Stirring Hotplates
Clamp
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    23
    Hours
    18
    Minutes
    30
    Seconds
    31
  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721
    Expires2025-05-01
    New Offer
    Save Now
Product Content Correction
Product Title
Fisherbrand™ Hotplate Stirrer Clamp > Hotplate/Stirrer Clamps