missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Gerbil Diet, Purified, Pellet

Purified gerbil diet, perfect for research and diet.

Brand:  MP Biomedicals 0296019720

 View more versions of this product

Product Code. 11412951

  • 17550.00 NOK / 20kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Purified gerbil diet, perfect for research and diet
  • Contains casein, methionine, inositol, gerbil salt mix, and MP Bio vitamin fortification mixture
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Single-use
20 kg
Room Temperature
0.3048 m
Animal Diet
Pellet
Non-sterile
1 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.