Learn More
Gentamycin Sulfate Solution (10mg Gentamycin Base Per ML), MP Biomedicals Product Code.: 11470435
compliance-icons compliance-icons
Gentamycin Sulfate Solution (10mg Gentamycin Base Per ML), MP Biomedicals
Product Code. 11470435 Shop All MP Biomedicals Products
10 x 10 mL
Click to view available options
Quantity:
10 x 10 mL
Product Code. 11470435
Brand: MP Biomedicals 0916760J8
Specifications
Type Gentamicin Sulfate
Quantity 10 x 10 mL
Chemical Name or Material Gentamycin Sulfate Solution
compliance-icons compliance-icons
Product Identifier
  • Gentamicin Sulfate Reagent Solution
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : gentamycin sulphate
Product Content Correction
Product Title
Gentamycin Sulfate Solution (10mg Gentamycin Base Per ML), MP Biomedicals >