missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genistein, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Specifications
Specifications
Color | White |
Quantity | 50 mg |
Formula Weight | 270.2 |
Physical Form | Free-flowing Powder |
Chemical Name or Material | Genistein |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Genistein, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction