Learn More
Thermo Scientific™ General Gas Chromatography Tubing and Tools
Use Thermo Scientific™ General Gas Chromatography Tubing and Tools to maintain and optimize your GC instrument.
Brand: Thermo Scientific™ 60201-318
82.87 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Use Thermo Scientific™ General Gas Chromatography Tubing and Tools to maintain and optimize your GC system and support your GC applications.
Specifications
Ceramic Column Cutter | |
Ceramic Column Cutter | |
GC | |
Gas Chromatography Columns |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%