Learn More
Thermo Scientific™ GeneBLAzer™ MCHR2-NFAT-bla CHO-K1 Cells
GeneBLAzer MCHR2-NFAT-bla CHO-K1 cells contain the human melanin-concentrating hormone receptor 2 (GPR145) stably integrated into the CellSensor NFAT-bla CHO-K1 parental cell line.
Brand: Thermo Scientific™ K1852
Description
GeneBLAzer MCHR2-NFAT-bla CHO-K1 cells contain the human melanin-concentrating hormone receptor 2 (GPR145) stably integrated into the CellSensor NFAT-bla CHO-K1 parental cell line. CellSensor NFAT-bla CHO-K1 cells contain a beta-lactamase reporter gene under control of a NFAT response element stably integrated into the cells. GeneBLAzer MCHR2-NFAT-bla CHO-K1 cells have been functionally validated for Z'-Factor and EC50 concentrations of MCH 6-17.
Specifications
NM_032503.2 | |
Dividing Cells | |
Fluorescence | |
G-Protein Coupled Receptor (GPCR) | |
GeneBLAzer™ | |
End Point | |
Dividing Cells | |
GeneBLAzer™ |
Cell-based beta lactamase reporter gene | |
GeneBLAzer™ MCHR2-NFAT-bla CHO-K1 cells are shipped on dry ice, 8 x 106 cells/vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. | |
FRET | |
Single Cell Clone Origin | |
1 Vial | |
GPR145, MCHR2 | |
CHO-K1 | |
GPCR |