Learn More
Cytiva Gel Filtration Calibration Kits
Shop All Cytiva Products
Click to view available options
MWCO:
43,000 to 669,000
6500 to 75,000
Unit Size:
1 set
Description
- Optimized range of proteins suited to high-resolution media and prepacked columns
- Each kit contains five vials of lyophilized proteins
- Blue Dextran 2000 included to determine the void fraction of the column
Specifications
Specifications
Quantity | Each |
For Use With (Equipment) | High-resolution media and prepacked column |
Product Type | Calibration Kit |
For Use With (Application) | Calibration of high-resolution gel filtration media and column such as Superdex™, Superose and Sephacryl |
Includes | 50mg Ovalbumin (43,000 Mr), 50mg Conalbumin (75,000 Mr), 50mg Aldolase (158,000 Mr), 15mg Ferritin (440,000 Mr), 50mg Thyroglobulin (669,000 Mr), 50mg Blue Dextran (2,000,000 Mr) |
MWCO | 43,000 to 669,000 |
Content And Storage | 2° to 8°C |
Product Content Correction
Product Title
Cytiva Gel Filtration Calibration Kits >