Learn More
Fisherbrand™ Gel Casting Gate
Product Code. 10134782
Maxi Plus Standard, Each
Click to view available options
For Use With (Equipment):
Maxi Plus Standard
Maxi Standard
Unit Size:
1 pair
Each
Product Code. 10134782
Brand: Fisherbrand™ HU25CG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With integral silicone seals to effectively seal tray.
  • Casting gates
  • Integral silicone seals effectively seal the tray without the need for tape
Specifications
For Use With (Equipment) Maxi Plus Standard
Type Gel Casting Gate
Includes Integral Silicone Seals
Product Content Correction
Product Title
Fisherbrand™ Gel Casting Gate >